missing translation for 'onlineSavingsMsg'
Learn More
Bromine liquid, 99.99+% (metals basis), Thermo Scientific Chemicals Product Code.: 15492967

Bromine liquid, 99.99+% (metals basis), Thermo Scientific Chemicals

Product Code. 15492967
5 mL
Click to view available options
Quantity:
5 mL
250 mL
This item is not returnable. View return policy

Product Code. 15492967

Brand: Thermo Scientific Alfa Aesar 087614.AA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]

Specifications

Color Red to Brown
Physical Form Liquid
Quantity 5 mL
Assay Percent Range 99.998% (metals basis)
IUPAC Name bromide
Percent Purity 99.99+%
Odor Unpleasant
Vapor Pressure 173mm Hg at 20°C
Chemical Name or Material Bromine liquid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromine liquid, 99.99+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.