missing translation for 'onlineSavingsMsg'
Learn More
Bromine, puriss. p.a., Honeywell Fluka™ Product Code.: 15621530

Bromine, puriss. p.a., Honeywell Fluka™

Product Code. 15621530 Shop All Honeywell Fluka Products
100 mL
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
This item is not returnable. View return policy

Product Code. 15621530

Brand: Honeywell-Fluka 16050100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
IUPAC Name bromide
SMILES [Br-]

Specifications

Quantity 100 mL
UN Number 1744
Formula Weight 159.81g/mol
Percent Purity ≥99% (RT)
Grade puriss. p.a.
Vapor Pressure 671mmHg (55°C), 175mmHg (20°C)
Density 3.119g/mL at 25°C; Vapor Density: 7.14 (vs air)
Chemical Name or Material Bromine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromine, puriss. p.a., Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.