Learn More
α-Bromo-p-xylene, 98%, Thermo Scientific Chemicals
£19.70 - £125.00
Chemical Identifiers
CAS | 104-81-4 |
---|---|
Molecular Formula | C8H9Br |
Molecular Weight (g/mol) | 185.06 |
MDL Number | MFCD00000181 |
InChI Key | WZRKSPFYXUXINF-UHFFFAOYSA-N |
Synonym | 4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene |
PubChem CID | 7721 |
IUPAC Name | 1-(bromomethyl)-4-methylbenzene |
SMILES | CC1=CC=C(C=C1)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10733203
|
Thermo Scientific Acros
107570050 |
5 g | Glass bottle |
£19.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10746131
|
Thermo Scientific Acros
107570250 |
25 g | Glass bottle |
£44.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10085943
|
Thermo Scientific
107571000 |
100 g | Glass bottle |
£125.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104-81-4 | |
185.06 | |
WZRKSPFYXUXINF-UHFFFAOYSA-N | |
7721 | |
CC1=CC=C(C=C1)CBr |
C8H9Br | |
MFCD00000181 | |
4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene | |
1-(bromomethyl)-4-methylbenzene |
Specifications
104-81-4 | |
100.0 | |
White to Yellow | |
97°C | |
97.5% min. (GC) | |
CH3C6H4CH2Br | |
05, 385 | |
4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene | |
WZRKSPFYXUXINF-UHFFFAOYSA-N | |
1-(bromomethyl)-4-methylbenzene | |
7721 | |
98% | |
α-Bromo-p-xylene |
97.5 | |
33.0°C to 37.0°C | |
218.0°C to 220.0°C | |
Authentic | |
C8H9Br | |
MFCD00000181 | |
15, 10288 | |
Solubility in water: practically insoluble. Other solubilities: soluble in chloroform and ether | |
CC1=CC=C(C=C1)CBr | |
185.06 | |
185.06 | |
Crystalline Mass |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-240-6
- a-Bromo-p-xylene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only