missing translation for 'onlineSavingsMsg'
Learn More

Bromo(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11335337
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11335337

Brand: Thermo Scientific Alfa Aesar 041002.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bromo(triphenylphosphine)gold(I) is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14243-65-3
Molecular Formula C18H15AuBrP
Molecular Weight (g/mol) 539.16
MDL Number MFCD00049267
InChI Key UIYUJCNGYSSFGP-UHFFFAOYSA-M
Synonym bromo triphenylphosphine gold i, bromoaurio triphenyl-??-phosphanyl
PubChem CID 12464140
IUPAC Name bromogold;triphenylphosphane
SMILES Br[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Physical Form Powder
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 539.17
Percent Purity 99.99%
Grade Premion
Assay (metals basis)
Chemical Name or Material Bromo(triphenylphosphine)gold(I)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.