missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoacetaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Density | 1.2800g/mL |
Boiling Point | 66.0°C to 67.0°C (18.0 mmHg) |
Flash Point | 61°C |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | BrCH2CH(OC2H5)2 |
Refractive Index | 1.4380 to 1.4400 |
Quantity | 500 mL |
Beilstein | 01,625 |
Specific Gravity | 1.28 |
Show More |
Safety and Handling
Product Identifier
- Bromoacetaldehyde diethyl acetal
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H301-Toxic if swallowed.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'