missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetaldehyde diethyl acetal, 97%

Product Code. 11480823
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11480823

Brand: Thermo Scientific Alfa Aesar A13276.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 2032-35-1 | C6H13BrO2 | 197.072 g/mol

Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ?-heterosubstituted acetaldehyde acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ɑ-heterosubstituted acetaldehyde acetals.

Solubility
It is immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Protect from frost, heat and sunlight. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Acids. Moisture Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2032-35-1
Molecular Formula C6H13BrO2
Molecular Weight (g/mol) 197.07
MDL Number MFCD00000214
InChI Key LILXDMFJXYAKMK-UHFFFAOYSA-N
Synonym bromoacetaldehyde diethyl acetal, bromoacetal, 2,2-diethoxyethyl bromide, ethane, 2-bromo-1,1-diethoxy, bromacetal, 1-bromo-2,2-diethoxyethane, diethyl bromoacetal, 2-bromoacetaldehyde diethyl acetal, 1,1-diethoxy-2-bromoethane, diethyl bromoacetaldehyde acetal
PubChem CID 74852
IUPAC Name 2-bromo-1,1-diethoxyethane
SMILES CCOC(CBr)OCC

Specifications

Density 1.27
Boiling Point 66°C to 67°C (18 mmHg)
Flash Point 61°C (142°F)
Linear Formula BrCH2CH(OCH2CH3)2
Refractive Index 1.439
Quantity 100 g
UN Number UN2810
Beilstein 635754
Sensitivity Moisture sensitive
Solubility Information It is immiscible with water.
Formula Weight 197.08
Percent Purity 97%
Chemical Name or Material Bromoacetaldehyde diethyl acetal
Show More Show Less
compliance-icons
Product Identifier
  • Bromoacetaldehyde diethyl acetal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Bromoacetaldehyde diethyl acetal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.