missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoacetaldehyde dimethyl acetal, 97+%, stab. with potassium carbonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1000 g
Description
2-Bromo-1,1-dimethoxyethane (Bromoacetaldehyde dimethyl acetal) was used in the synthesis of 2,3-O-acetal via reaction with 2,3-diol in the presence of 10-camphorsulphonic acid (CSA). 2-bromo-1,1-dimethoxyethane was used to react with 4-hydroxylbenzaldehyde, and finally aldehyde substituted prodrug was generated by TFA-mediated deacetalization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-1,1-dimethoxyethane (Bromoacetaldehyde dimethyl acetal) was used in the synthesis of 2,3-O-acetal via reaction with 2,3-diol in the presence of 10-camphorsulphonic acid (CSA). 2-bromo-1,1-dimethoxyethane was used to react with 4-hydroxylbenzaldehyde, and finally aldehyde substituted prodrug was generated by TFA-mediated deacetalization.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
2-Bromo-1,1-dimethoxyethane (Bromoacetaldehyde dimethyl acetal) was used in the synthesis of 2,3-O-acetal via reaction with 2,3-diol in the presence of 10-camphorsulphonic acid (CSA). 2-bromo-1,1-dimethoxyethane was used to react with 4-hydroxylbenzaldehyde, and finally aldehyde substituted prodrug was generated by TFA-mediated deacetalization.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.435 |
Boiling Point | 150°C |
Flash Point | 54°C (129°F) |
Linear Formula | BrCH2CH(OCH3)2 |
Refractive Index | 1.445 |
Quantity | 25 g |
UN Number | UN2929 |
Beilstein | 1280132 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'