Learn More
Bromoacetic acid, 99%, Thermo Scientific Chemicals
£34.67 - £441.20
Chemical Identifiers
CAS | 79-08-3 |
---|---|
Molecular Formula | C2H3BrO2 |
Molecular Weight (g/mol) | 138.95 |
MDL Number | MFCD00002678 |
InChI Key | KDPAWGWELVVRCH-UHFFFAOYSA-N |
Synonym | bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo |
PubChem CID | 6227 |
IUPAC Name | 2-bromoacetic acid |
SMILES | C(C(=O)O)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10316360
|
Thermo Scientific Acros
106572500 |
250 g | Glass bottle |
£34.67
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10112420
|
Thermo Scientific Acros
106570010 |
1 kg | Glass bottle |
£88.14
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10216883
|
Thermo Scientific Acros
106570025 |
2.5 kg | Glass bottle |
£166.83
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13498298
|
Thermo Scientific Acros
106570100 |
10 kg | Plastic Pail |
£441.20
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-08-3 | |
138.95 | |
KDPAWGWELVVRCH-UHFFFAOYSA-N | |
6227 | |
C(C(=O)O)Br |
C2H3BrO2 | |
MFCD00002678 | |
bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo | |
2-bromoacetic acid |
Specifications
79-08-3 | |
White to Yellow | |
208.0°C | |
Authentic | |
C2H3BrO2 | |
MFCD00002678 | |
15,1407 | |
bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo | |
KDPAWGWELVVRCH-UHFFFAOYSA-N | |
2-bromoacetic acid | |
6227 | |
99% | |
Bromoacetic acid |
49.0°C to 51.0°C | |
1.9300g/mL | |
113°C | |
99% | |
BrCH2CO2H | |
02,213 | |
1.93 | |
Solubility in water: 900g/100mL (25°C). Other solubilities:g/100g(25°C): methanol-900,ether-735,benzene-308,carbon tetrachloride-168,soluble in acetone and alcohol,practically insoluble in n-heptane | |
C(C(=O)O)Br | |
138.95 | |
138.95 | |
Crystalline Mass or Crystalline Powder, Flakes, Crystals and/or Chunks |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
May cause an allergic skin reaction.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-175-8
- Bromoacetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Skin sensitiser Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.