Learn More
Bromoacetonitrile, 97%, Thermo Scientific Chemicals
£20.90 - £192.00
Chemical Identifiers
CAS | 590-17-0 |
---|---|
Molecular Formula | C2H2BrN |
Molecular Weight (g/mol) | 119.95 |
MDL Number | MFCD00001884 |
InChI Key | REXUYBKPWIPONM-UHFFFAOYSA-N |
Synonym | bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile |
PubChem CID | 11534 |
IUPAC Name | 2-bromoacetonitrile |
SMILES | BrCC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10451071
|
Thermo Scientific Acros
221860050 |
5 g | Glass bottle |
£20.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10164750
|
Thermo Scientific Acros
221860250 |
25 g | Glass bottle |
£61.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10246370
|
Thermo Scientific Acros
221861000 |
100 g | Glass bottle |
£192.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
590-17-0 | |
119.95 | |
REXUYBKPWIPONM-UHFFFAOYSA-N | |
11534 | |
BrCC#N |
C2H2BrN | |
MFCD00001884 | |
bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile | |
2-bromoacetonitrile |
Specifications
590-17-0 | |
1.7200g/mL | |
>110°C | |
96% min. (GC) | |
BrCH2CN | |
MFCD00001884 | |
13,197 | |
bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile | |
BrCC#N | |
119.95 | |
119.95 | |
Liquid |
Colorless to Yellow | |
148°C to 150°C | |
Authentic | |
C2H2BrN | |
1.4786 to 1.4806 | |
02, 216 | |
1.72 | |
REXUYBKPWIPONM-UHFFFAOYSA-N | |
2-bromoacetonitrile | |
11534 | |
97% | |
Bromoacetonitrile, 97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-672-1
missing translation for 'rtecsNumber' : AL7970000
- Bromoacetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only