Learn More
Bromobenzene, 99%, pure, Thermo Scientific Chemicals
£34.37 - £534.66
Chemical Identifiers
CAS | 108-86-1 |
---|---|
Molecular Formula | C6H5Br |
Molecular Weight (g/mol) | 157.01 |
MDL Number | MFCD00000055 |
InChI Key | QARVLSVVCXYDNA-UHFFFAOYSA-N |
Synonym | benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47 |
PubChem CID | 7961 |
ChEBI | CHEBI:3179 |
IUPAC Name | bromobenzene |
SMILES | BrC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10599810
|
Thermo Scientific Acros
106682500 |
250 mL | Glass bottle |
£34.37
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10589810
|
Thermo Scientific Acros
106680010 |
1 L | Glass bottle |
£156.78
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11350320
|
Thermo Scientific Acros
106680025 |
2.5 L | Glass bottle |
£178.89
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13408308
|
Thermo Scientific Acros
106680100 |
10 L | Plastic Drum |
£534.66
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
108-86-1 | |
157.01 | |
QARVLSVVCXYDNA-UHFFFAOYSA-N | |
7961 | |
bromobenzene |
C6H5Br | |
MFCD00000055 | |
benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47 | |
CHEBI:3179 | |
BrC1=CC=CC=C1 |
Especificaciones
108-86-1 | |
Colorless to Yellow | |
156.0°C | |
Authentic | |
C6H5Br | |
1.5580 to 1.5600 | |
05,206 | |
1.49 | |
Solubility in water: practically insoluble. Other solubilities: 10.4g/100g alcohol (25 c),71.3g/100g ether (25 c),miscible with chloroform and benzene | |
BrC1=CC=CC=C1 | |
157.01 | |
CHEBI:3179 | |
157.01 | |
Pure | |
Bromobenzene |
-31.0°C | |
1.4900g/mL | |
51°C | |
98.5% min. (GC) | |
C6H5Br | |
MFCD00000055 | |
15,1413 | |
benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47 | |
QARVLSVVCXYDNA-UHFFFAOYSA-N | |
bromobenzene | |
7961 | |
1.124 mPa.s (20°C) | |
99% | |
Liquid |
Seguridad y manipulación
GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-623-8
- Bromobenzene
Palabra de advertencia
- Warning
Clasificación
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
Indicaciones de peligro
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H226-Flammable liquid and vapour.
Consejos de prudencia
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362-Take off contaminated clothing.
RUO – Research Use Only