missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
Descripción
The labeled version of bromobenzene , used as a general reagent in palladium-catalyzed reaction and in the synthesize of Grignard reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
Physical Form | Liquid |
Density | 1.55 g/mL |
Flash Point | 51°C (124°F) |
Odor | Aromatic |
Refractive Index | 1.557 |
Quantity | 10 g |
Boiling Point | 53°C (23mmHg) |
UN Number | UN2514 |
Solubility Information | Insoluble in water. |
IUPAC Name | 1-bromo-2,3,4,5,6-pentadeuteriobenzene |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'