missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Bromocinnamaldehyde, 98%, Thermo Scientific™
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Specifications
Melting Point | 68.0°C to 72.0°C |
Color | Yellow to Ochre |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C9H7BrO |
Linear Formula | C6H5CH=C(Br)CHO |
Quantity | 250 g |
Beilstein | 07,358 |
Molecular Weight (g/mol) | 211.06 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- a-Bromocinnamaldehyde
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'