Learn More
Description
Bromocresol Green is a pH sensitive indicator used in the growth medium for microorganisms and titrations. It is useful in various colorimetric detection studies and as an actively tracking dye for DNA agarose gel electrophoresis. It acts as an inhibitor of prostaglandin E2 transport protein. It is also used in TLC for visualizing compounds containing functional groups having pKa less than 5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromocresol Green is a pH sensitive indicator used in the growth medium for microorganisms and titrations. It is useful in various colorimetric detection studies and as an actively tracking dye for DNA agarose gel electrophoresis. It acts as an inhibitor of prostaglandin E2 transport protein. It is also used in TLC for visualizing compounds containing functional groups having pKa less than 5.
Solubility
Soluble in benzene, ethanol and diethyl ether. Sparingly soluble in water.
Notes
Transition interval: pH 3.8 (yellow) to pH 5.4 (blue).Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | ∼225°C (decomposition) |
Beilstein | 372527 |
Merck Index | 14,1386 |
Solubility Information | Soluble in benzene,ethanol and diethyl ether. Sparingly soluble in water. |
Formula Weight | 698.02 |
Quantity | 5 g |
Chemical Name or Material | Bromocresol Green |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253473 | Certificate of Analysis | 07/01/2025 | A17090.14,A17090.06 |
10248004 | Certificate of Analysis | 02/10/2023 | A17090.14,A17090.06 |
10242333 | Certificate of Analysis | 25/08/2022 | A17090.14,A17090.06 |
10112509 | Certificate of Analysis | 23/11/2021 | A17090.14,A17090.06 |
10234797 | Certificate of Analysis | 23/11/2021 | A17090.14,A17090.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.