missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromocresol Green, ACS reagent
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Melting Point | 225.0°C |
Clarity | Passes Test |
Merck Index | 15, 1393 |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in benzene, ethanol (30g/L), very soluble in diethyl ether and ethylacetate |
Infrared Spectrum | Authentic |
Formula Weight | 698.01 |
Grade | ACS Reagent |
Packaging | Glass bottle |
Visual Transition Interval | from pH 3.8 (yellow) to pH 5.4 (blue) |
Color | Beige to Brown |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'