missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Indicator 0.04% Solution, Reagecon
BIOPHARMA

High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.

Brand:  Reagecon BRCG05


Product Code. 11533212

  • £55.20 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • For manual titrations; indicator titrant or analyte change color and this color change are by far the most important method of end point detection.
  • This high-quality indicator solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications

Specifications

0.0004
Amber Glass Winchester Bottle GL45
500 mL
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BROMOCRESOL GREEN INDICATOR

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : propanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers