missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromocresol Green sodium salt, 0.04% w/v aq. soln.
£25.53 - £42.41
Chemical Identifiers
CAS | 62625-32-5 |
---|---|
MDL Number | MFCD00148898 |
Synonym | Bromocresol Green, water soluble |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11328967
|
Thermo Scientific Chemicals
038696.AE |
100 mL |
£25.53
100mL |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
11460151
|
Thermo Scientific Chemicals
038696.AP |
500 mL |
£42.41
500mL |
In Stock
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62625-32-5 | |
MFCD00148898 | |
Bromocresol Green, water soluble |
Specifications
62625-32-5 | |
MFCD00148898 | |
720.01 | |
Liquid |
0.04% w/v aq. soln. | |
Bromocresol Green, water soluble | |
Green | |
Bromocresol Green sodium salt |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only