missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, Water Soluble, Indicator

£24.82 - £50.75

Chemical Identifiers

CAS 67763-24-0
Molecular Formula C21H13Br4NaO5S
Molecular Weight (g/mol) 720.00
MDL Number MFCD00148898
InChI Key HEFSGAHJDGZCHA-UHFFFAOYNA-M
Synonym 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein sodium salt
PubChem CID 23675901
IUPAC Name sodium;2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenolate
SMILES [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10153200
View Documents
Thermo Scientific Chemicals
151320050
5 g Glass bottle
£24.82
5g
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
 
10337660
View Documents
Thermo Scientific Chemicals
151320250
25 g Glass bottle
£50.75
25g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

67763-24-0
720.00
HEFSGAHJDGZCHA-UHFFFAOYNA-M
23675901
[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
C21H13Br4NaO5S
MFCD00148898
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein sodium salt
sodium;2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenolate

Specifications

67763-24-0
C21H13Br4NaO5S
3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein sodium salt
Authentic
sodium;2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenolate
23675901
Extra Pure
from pH 3.8 (yellow) to pH 5.4 (blue)
Crystalline Powder
(0.1% in water) clear
MFCD00148898
HEFSGAHJDGZCHA-UHFFFAOYNA-M
[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
720.00
720
Complies
Black/Green to Brown
Bromocresol Green, sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only