missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclopentane, 98+%, Thermo Scientific Chemicals

£28.54 - £83.42

Chemical Identifiers

CAS 137-43-9
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00001359
InChI Key BRTFVKHPEHKBQF-UHFFFAOYSA-N
Synonym cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
PubChem CID 8728
IUPAC Name bromocyclopentane
SMILES C1CCC(C1)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11482807
View Documents
Thermo Scientific Alfa Aesar
A15455.22
100 g
£28.54
100g
Please to purchase this item. Need a web account? Register with us today!
11492807
View Documents
Thermo Scientific Alfa Aesar
A15455.36
500 g
£83.42
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bromocyclopentane is a precursor used in the preparation of cyclopentene and cyclopentanol. It is a fatty bromide, which is used as a solvent. It is used as an intermediate for the preparation of surfactant and pharmaceuticals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocyclopentane is a precursor used in the preparation of cyclopentene and cyclopentanol. It is a fatty bromide, which is used as a solvent. It is used as an intermediate for the preparation of surfactant and pharmaceuticals and other organic compounds.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

137-43-9
149.031
BRTFVKHPEHKBQF-UHFFFAOYSA-N
8728
C1CCC(C1)Br
C5H9Br
MFCD00001359
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
bromocyclopentane

Specifications

137-43-9
137°C to 139°C
98%
1.4895
UN1993
cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
BRTFVKHPEHKBQF-UHFFFAOYSA-N
bromocyclopentane
8728
≥98%
Bromocyclopentane
1.390
42°C (107°F)
C5H9Br
MFCD00001359
1209256
Immiscible with water.
C1CCC(C1)Br
149.031
149.04
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 205-294-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only