Learn More
Bromoethane, ≥99%, extra pure, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 74-96-4 |
---|---|
Molecular Formula | C2H5Br |
Molecular Weight (g/mol) | 108.966 |
MDL Number | MFCD00000232 |
InChI Key | RDHPKYGYEGBMSE-UHFFFAOYSA-N |
Synonym | ethyl bromide, ethane, bromo, 1-bromoethane, monobromoethane, bromic ether, hydrobromic ether, bromure d'ethyle, ethylbromide, etylu bromek, halon 2001 |
PubChem CID | 6332 |
IUPAC Name | bromoethane |
SMILES | CCBr |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
74-96-4 | |
108.966 | |
RDHPKYGYEGBMSE-UHFFFAOYSA-N | |
6332 | |
CCBr |
C2H5Br | |
MFCD00000232 | |
ethyl bromide, ethane, bromo, 1-bromoethane, monobromoethane, bromic ether, hydrobromic ether, bromure d'ethyle, ethylbromide, etylu bromek, halon 2001 | |
bromoethane |
Specifications
0.0005 % max. (as HBr) | |
Brown to White | |
99+% | |
C2H5Br | |
MFCD00000232 | |
RDHPKYGYEGBMSE-UHFFFAOYSA-N | |
bromoethane | |
6332 | |
≥99.0% (GC) | |
Crystals or Powder |
74-96-4 | |
Authentic | |
C2H5Br | |
1.4230 to 1.4250 (20°C, 589nm) | |
ethyl bromide, ethane, bromo, 1-bromoethane, monobromoethane, bromic ether, hydrobromic ether, bromure d'ethyle, ethylbromide, etylu bromek, halon 2001 | |
CCBr | |
108.966 | |
108.97 | |
Extra Pure | |
Bromoethane, 99+% |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-825-8
missing translation for 'rtecsNumber' : KH6475000
missing translation for 'tsca' : TSCA
RUO – Research Use Only