missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoform, 96%, stab. with ethanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.
Solubility
Slightly soluble in water.
Notes
Light-sensitive. A lachrymator. Incompatible with chemically active metals and strong bases.
Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.
Solubility
Slightly soluble in water.
Notes
Light-sensitive. A lachrymator. Incompatible with chemically active metals and strong bases.
Specifications
Specifications
Melting Point | 8°C to 9°C |
Density | 2.81 |
Boiling Point | 148°C to 150°C |
Flash Point | None |
Refractive Index | 1.585 |
Quantity | 1000 g |
UN Number | UN2515 |
Beilstein | 1731048 |
Sensitivity | Light sensitive |
Merck Index | 14,1420 |
Show More |
Safety and Handling
Product Identifier
- Bromoform, 97%, stab. with ethanol
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : Bromoform, ethanol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'