missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, 96%, stabilized, Thermo Scientific Chemicals

Product Code. 10573244
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 75-25-2, 64-17-5
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.73
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br

Especificaciones

Melting Point 8.0°C
Color Colorless to Orange-Yellow
Density 2.6300g/mL
Boiling Point 150.0°C to 151.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic drum
Linear Formula CHBr3
Quantity 2.5 L
Beilstein 01,68
Fieser 12,74
Merck Index 15,143
Specific Gravity 2.63
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,benzene,chloroform,ether,,petroleum ether,acetone and oils
Stabilizer 3 to 4 wt% ethanol
Formula Weight 252.73
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Bromoform
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'