Learn More
Bromomaleic anhydride, 97%, Thermo Scientific Chemicals
£64.30 - £219.00
Chemical Identifiers
CAS | 5926-51-2 |
---|---|
Molecular Formula | C4HBrO3 |
Molecular Weight (g/mol) | 176.95 |
MDL Number | MFCD00005519 |
InChI Key | YPRMWCKXOZFJGF-UHFFFAOYSA-N |
Synonym | bromomaleic anhydride, 2,5-furandione, 3-bromo, 3-bromo-2,5-furandione, bromomaleicanhydride, monobromomaleic anhydride, 3-bromo-2,5-furanedione, 2,5-furandione,3-bromo, 2-bromomaleic acid anhydride, 3-bromo-2,5-furandione # |
PubChem CID | 80027 |
IUPAC Name | 3-bromofuran-2,5-dione |
SMILES | BrC1=CC(=O)OC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10693995
|
Thermo Scientific Acros
147450100 |
10 g | Glass bottle |
£64.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10122834
|
Thermo Scientific Acros
147450500 |
50 g | Glass bottle |
£219.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5926-51-2 | |
176.95 | |
YPRMWCKXOZFJGF-UHFFFAOYSA-N | |
80027 | |
BrC1=CC(=O)OC1=O |
C4HBrO3 | |
MFCD00005519 | |
bromomaleic anhydride, 2,5-furandione, 3-bromo, 3-bromo-2,5-furandione, bromomaleicanhydride, monobromomaleic anhydride, 3-bromo-2,5-furanedione, 2,5-furandione,3-bromo, 2-bromomaleic acid anhydride, 3-bromo-2,5-furandione # | |
3-bromofuran-2,5-dione |
Specifications
5926-51-2 | |
215.0°C | |
96% min. (GC) | |
1.54 | |
17, 435 | |
bromomaleic anhydride, 2,5-furandione, 3-bromo, 3-bromo-2,5-furandione, bromomaleicanhydride, monobromomaleic anhydride, 3-bromo-2,5-furanedione, 2,5-furandione,3-bromo, 2-bromomaleic acid anhydride, 3-bromo-2,5-furandione # | |
BrC1=CC(=O)OC1=O | |
176.95 | |
176.95 | |
Bromomaleic anhydride |
1.9050g/mL | |
113°C | |
C4HBrO3 | |
MFCD00005519 | |
1.905 | |
YPRMWCKXOZFJGF-UHFFFAOYSA-N | |
3-bromofuran-2,5-dione | |
80027 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 227-659-9
- Bromomaleic anhydride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only