Learn More
(Bromomethyl)cyclohexane, 98%, Thermo Scientific Chemicals
£21.20 - £56.00
Chemical Identifiers
CAS | 2550-36-9 |
---|---|
Molecular Formula | C7H13Br |
Molecular Weight (g/mol) | 177.09 |
MDL Number | MFCD00001509 |
InChI Key | UUWSLBWDFJMSFP-UHFFFAOYSA-N |
Synonym | bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane |
PubChem CID | 137636 |
IUPAC Name | bromomethylcyclohexane |
SMILES | BrCC1CCCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317726
|
Thermo Scientific Alfa Aesar
L11343.06 |
5 g |
£21.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327726
|
Thermo Scientific Alfa Aesar
L11343.14 |
25 g |
£56.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(Bromomethyl)cyclohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Bromomethyl)cyclohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
Chemical Identifiers
2550-36-9 | |
177.09 | |
UUWSLBWDFJMSFP-UHFFFAOYSA-N | |
137636 | |
BrCC1CCCCC1 |
C7H13Br | |
MFCD00001509 | |
bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane | |
bromomethylcyclohexane |
Specifications
2550-36-9 | |
99°C to 100°C (40 mmHg) | |
C7H13Br | |
MFCD00001509 | |
bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane | |
UUWSLBWDFJMSFP-UHFFFAOYSA-N | |
bromomethylcyclohexane | |
137636 | |
98% |
1.269 | |
68°C (154°F) | |
1.4935 | |
635738 | |
Insoluble in water. | |
BrCC1CCCCC1 | |
177.09 | |
177.09 | |
(Bromomethyl)cyclohexane |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only