Learn More
(Bromomethyl)cyclopropane, 97%, Thermo Scientific Chemicals
£37.40 - £425.00
Chemical Identifiers
CAS | 7051-34-5 |
---|---|
Molecular Formula | C4H7Br |
Molecular Weight (g/mol) | 135.00 |
MDL Number | MFCD00001306 |
InChI Key | AEILLAXRDHDKDY-UHFFFAOYSA-N |
Synonym | bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide |
PubChem CID | 81503 |
SMILES | BrCC1CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394464
|
Thermo Scientific Alfa Aesar
L01122.03 |
1 g |
£37.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304474
|
Thermo Scientific Alfa Aesar
L01122.06 |
5 g |
£114.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314474
|
Thermo Scientific Alfa Aesar
L01122.14 |
25 g |
£425.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(Bromomethyl)cyclopropane is used in the synthesis of 1,4-dienes by iron-catalyzed cross-coupling with alkenyl Grignard reagents. It has been used as a synthetic building block for the introduction of the cyclopropylmethyl group.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Bromomethyl)cyclopropane is used in the synthesis of 1,4-dienes by iron-catalyzed cross-coupling with alkenyl Grignard reagents. It has been used as a synthetic building block for the introduction of the cyclopropylmethyl group.
Solubility
Not miscible in water.
Notes
Protect from heat. Store away from oxidizing agents. Incompatible with strong bases, strong oxidizing agents.
Chemical Identifiers
7051-34-5 | |
135.00 | |
AEILLAXRDHDKDY-UHFFFAOYSA-N | |
81503 |
C4H7Br | |
MFCD00001306 | |
bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide | |
BrCC1CC1 |
Specifications
7051-34-5 | |
111°C to 112°C | |
C4H7Br | |
MFCD00001306 | |
605296 | |
Not miscible in water. | |
BrCC1CC1 | |
135.00 | |
135 | |
(Bromomethyl)cyclopropane |
1.41 | |
41°C (105°F) | |
1.477 | |
UN1993 | |
bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide | |
AEILLAXRDHDKDY-UHFFFAOYSA-N | |
(bromomethyl)cyclopropane | |
81503 | |
97% |
Safety and Handling
GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 230-331-8
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only