missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromopentamethylbenzene, 98%, Thermo Scientific Chemicals
£38.90 - £153.00
Chemical Identifiers
CAS | 5153-40-2 |
---|---|
Molecular Formula | C11H15Br |
Molecular Weight (g/mol) | 227.15 |
MDL Number | MFCD00013523 |
InChI Key | XPDQRULPGCFCLX-UHFFFAOYSA-N |
Synonym | bromopentamethylbenzene, benzene, bromopentamethyl, pentamethylbromobenzene, 1-bromopentamethylbenzene, 1-bromo-2,3,4,5,6-pentamethyl-benzene, brompentamethylbenzol, maybridge1_002545, ksc273s6f, acmc-1b227, 5-bromo-1,2,3,4,6-pentamethylbenzene |
PubChem CID | 78831 |
IUPAC Name | 1-bromo-2,3,4,5,6-pentamethylbenzene |
SMILES | CC1=C(C)C(C)=C(Br)C(C)=C1C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397705
|
Thermo Scientific Alfa Aesar
L02451.06 |
5 g |
£38.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307715
|
Thermo Scientific Alfa Aesar
L02451.14 |
25 g |
£153.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
5153-40-2 | |
227.15 | |
XPDQRULPGCFCLX-UHFFFAOYSA-N | |
78831 | |
CC1=C(C)C(C)=C(Br)C(C)=C1C |
C11H15Br | |
MFCD00013523 | |
bromopentamethylbenzene, benzene, bromopentamethyl, pentamethylbromobenzene, 1-bromopentamethylbenzene, 1-bromo-2,3,4,5,6-pentamethyl-benzene, brompentamethylbenzol, maybridge1_002545, ksc273s6f, acmc-1b227, 5-bromo-1,2,3,4,6-pentamethylbenzene | |
1-bromo-2,3,4,5,6-pentamethylbenzene |
Specifications
5153-40-2 | |
C11H15Br | |
1863555 | |
Insoluble in water. | |
CC1=C(C)C(C)=C(Br)C(C)=C1C | |
227.15 | |
227.15 | |
Bromopentamethylbenzene |
161°C to 162°C | |
MFCD00013523 | |
bromopentamethylbenzene, benzene, bromopentamethyl, pentamethylbromobenzene, 1-bromopentamethylbenzene, 1-bromo-2,3,4,5,6-pentamethyl-benzene, brompentamethylbenzol, maybridge1_002545, ksc273s6f, acmc-1b227, 5-bromo-1,2,3,4,6-pentamethylbenzene | |
XPDQRULPGCFCLX-UHFFFAOYSA-N | |
1-bromo-2,3,4,5,6-pentamethylbenzene | |
78831 | |
≥98% |
Safety and Handling
missing translation for 'einecsNumber' : 225-926-4
missing translation for 'tsca' : No
RUO – Research Use Only