missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Bromophenol Blue Product Code.: 10015540

Thermo Scientific Chemicals Bromophenol Blue

Product Code. 10015540
5 g, Glass bottle
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10015540

Brand: Thermo Scientific Chemicals 403140050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 279.0°C
Clarity Passes Test
Beilstein 19, I, 649
Merck Index 15, 1454
Infrared Spectrum Authentic
Formula Weight 669.96
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 3.0 (yellow) to pH 4.6 (blue)
Color Orange/Pink to Red or Purple
Quantity 5 g
Physical Form Powder
Chemical Name or Material Bromophenol Blue, Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465634 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog Number403140500,403140100,403140050
Lot NumberA0393461 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number403140500,403140100,403140050
Lot NumberA0411358 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number403140500,403140100,403140050
Lot NumberA0445070 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog Number403140500,403140100,403140050
Lot NumberA0353913 Certificate TypeCertificate of Analysis Date15/12/2020 Catalog Number403140500,403140100,403140050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Bromophenol Blue >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.