Learn More
Bromophenol Blue Indicator 0.04% Solution, Reagecon
High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
Brand: Reagecon BRBP05
Code : NEW
Additional Details : CAS Number : 67-63-0 Weight : 0.40000kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- For manual titrations; indicator titrant or analyte change color and this color change are by far the most important method of end point detection.
- This high-quality indicator solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
0.0004 | |
HDPE Bottle GL45 | |
500 mL | |
Liquid |
Safety and Handling
- bromophenol blue 0.04% indicator
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol