missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, Indicator, Reag. Ph. Eur., Honeywell Fluka™

Product Code. 15603840
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 3′,3′′,5′,5′′-Tetrabromophenolsulfonephthalein, Bromophenol Blue (BPB), Bromphenol Blue Sultone Form
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

CAS Max % 100.0000%
Linear Formula C19H10Br4O5S
Beilstein 61698
Formula Weight 669.96
Grade Ph. Eur.
Packaging Glass Bottle
Quantity 25 g
Chemical Name or Material Bromophenol Blue, Reag. Ph. Eur., indicator
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'