missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue, pure, 0.1% in water, Thermo Scientific Chemicals
£52.30 - £159.00
Specifications
Chemical Name or Material | Bromophenol Blue |
---|---|
CAS | 115-39-9, 7732-18-5 |
Concentration | about 0.1% (by UV) |
Molecular Formula | C19H10Br4O5S |
Synonym | 3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5' |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10187220
|
Thermo Scientific Acros
403150250 |
25 mL | Glass bottle |
£52.30
25mL |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
11956831
|
Thermo Scientific Acros
403151000 |
100 mL | Glass bottle |
£159.00
100mL |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
Bromophenol Blue | |
about 0.1% (by UV) | |
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5' | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br | |
669.96 | |
CHEBI:59424 | |
Pure | |
1 |
115-39-9, 7732-18-5 | |
C19H10Br4O5S | |
UDSAIICHUKSCKT-UHFFFAOYSA-N | |
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol | |
8272 | |
669.96 | |
Glass bottle | |
1.0000g/mL |
RUO – Research Use Only