missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS reagent
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Melting Point | >300.0°C |
Beilstein | 19, I, 649 |
SMILES | C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+] |
Formula Weight | 691.94 |
Grade | ACS Reagent |
Packaging | Glass bottle |
Quantity | 50 g |
Chemical Name or Material | Bromophenol Blue, sodium salt, Pure |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'