missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromothymol Blue, Indicator
£72.76 - £872.34
Chemical Identifiers
CAS | 76-59-5 |
---|---|
Molecular Formula | C27H28Br2O5S |
Molecular Weight (g/mol) | 624.384 |
MDL Number | MFCD00005872 |
InChI Key | NUHCTOLBWMJMLX-UHFFFAOYSA-N |
Synonym | Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB |
PubChem CID | 6450 |
ChEBI | CHEBI:86155 |
IUPAC Name | 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol |
SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10397750
|
Thermo Scientific Chemicals
151360250 |
25 g | Glass bottle |
£72.76
25g |
In Stock
Log in to see stock. |
||||
10307760
|
Thermo Scientific Chemicals
151361000 |
100 g | Glass bottle |
£235.17
100g |
In Stock
Log in to see stock. |
||||
10794284
|
Thermo Scientific Chemicals
151365000 |
500 g | Glass bottle |
£872.34
500g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
76-59-5 | |
624.384 | |
NUHCTOLBWMJMLX-UHFFFAOYSA-N | |
6450 | |
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol |
C27H28Br2O5S | |
MFCD00005872 | |
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB | |
CHEBI:86155 | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br |
Specifications
204.0°C | |
C27H28Br2O5S | |
15, 1455 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, alkaline solution | |
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br | |
624.384 | |
CHEBI:86155 | |
Pure |
76-59-5 | |
MFCD00005872 | |
Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB | |
NUHCTOLBWMJMLX-UHFFFAOYSA-N | |
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol | |
6450 | |
624.38 | |
Bromothymol Blue |
Safety and Handling
missing translation for 'einecsNumber' : 200-971-2
missing translation for 'rtecsNumber' : SJ7450000
missing translation for 'tsca' : TSCA
RUO – Research Use Only