missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue, sodium salt, pure, water soluble, indicator

£78.39 - £217.08

Chemische Identifikatoren

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
Synonym Bromthymol Blue, sodium salt, 3', 3''-Dibromothymolsulfonephthalein, sodium salt, BTB
PubChem CID 102183223
IUPAC Name sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Quantity Packaging Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Packaging Preis Menge & Verfügbarkeit  
10487360
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
25 g Glass bottle
£78.39
25g
In Stock
to see stock.
Zum Warenkorb hinzufügen
 
10711555
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
100 g Glass bottle
£217.08
100g
In Stock
to see stock.
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Spezifikation

Chemische Identifikatoren

34722-90-2
646.37
NMKFVGALBGZKGW-FKWCIMQXSA-M
102183223
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
C27H27Br2NaO5S
MFCD00077263,MFCD00077263,MFCD00077263
Bromthymol Blue, sodium salt, 3', 3''-Dibromothymolsulfonephthalein, sodium salt, BTB
sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate

Spezifikation

34722-90-2
MFCD00077263,MFCD00077263,MFCD00077263
Bromthymol Blue, sodium salt, 3', 3''-Dibromothymolsulfonephthalein, sodium salt, BTB
NMKFVGALBGZKGW-FKWCIMQXSA-M
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
646.37
646.35
Complies
Brown/Orange to Green
Bromothymol Blue, sodium salt
C27H27Br2NaO5S
15, 1455
Solubility in water: soluble.
Authentic
sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate
102183223
Pure
from pH 6.0 (yellow) to pH 7.6 (blue)
Powder
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

missing translation for 'einecsNumber' : 252-169-7

missing translation for 'tsca' : TSCA

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only