missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue Solution R1, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
1 Ea.
Unit Size:
100mL
Description
- Product Identifier: BROMOTHYMOL BLUE SOLUTION R1
- Signal Word: Danger
- Hazard Category: Flammable liquid Category 2, Serious eye damage/eye irritation Category 2
- Hazard Statement: H225-Highly flammable liquid and vapour, H319-Causes serious eye irritation.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241- Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
MIXTURE LIST - Contains: ethanol
Specifications
Specifications
| Color | Blue |
| Quantity | 1 Ea. |
Safety and Handling
Product Identifier
- BROMOTHYMOL BLUE SOLUTION R1
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction