missing translation for 'onlineSavingsMsg'
Learn More
Bromotrimethylsilane, 98%, AcroSeal™ Product Code.: 10329847

Bromotrimethylsilane, 98%, AcroSeal™

Product Code. 10329847
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy

Product Code. 10329847

Brand: Thermo Scientific Acros 430751000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2857-97-8 | C3H9BrSi | 153.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 2857-97-8
Melting Point -43.0°C
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 79.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging AcroSeal™ Glass bottle
Molecular Formula C3H9BrSi
Linear Formula (CH3)3SiBr
Refractive Index 1.4200 to 1.4250
MDL Number MFCD00000048
Quantity 100 mL
Specific Gravity 1.16
Synonym trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
Solubility Information Solubility in water: decomposes.
InChI Key IYYIVELXUANFED-UHFFFAOYSA-N
SMILES C[Si](C)(C)Br
IUPAC Name bromo(trimethyl)silane
Molecular Weight (g/mol) 153.1
PubChem CID 76113
Formula Weight 153.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bromotrimethylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460428 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number430751000
Lot NumberA0456627 Certificate TypeCertificate of Analysis Date26/09/2023 Catalog Number430751000
Lot NumberA0434430 Certificate TypeCertificate of Analysis Date07/09/2023 Catalog Number430751000
Lot NumberA0442973 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number430751000
Lot NumberA0395830 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number430751000
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromotrimethylsilane, 98%, AcroSeal™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.