missing translation for 'onlineSavingsMsg'
Learn More

Bromotriphenylmethane, 98%, Thermo Scientific Chemicals

Product Code. 11415293
Click to view available options
Quantity:
10 g
50 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is considered as a more reactive reagent than Chlorotriphenyl­methane, for the preparation of trityl esters. It is also used in one-pot conversion of amino acids to their N-trityl derivatives involving N,O-ditritylation and selective methanolysis of the trityl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is considered as a more reactive reagent than Chlorotriphenyl­methane, for the preparation of trityl esters. It is also used in one-pot conversion of amino acids to their N-trityl derivatives involving N,O-ditritylation and selective methanolysis of the trityl ester.

Solubility
Solubility in hot acetic acid is almost transparent.

Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-43-0
Molecular Formula C19H15Br
Molecular Weight (g/mol) 323.233
MDL Number MFCD00000120
InChI Key NZHXEWZGTQSYJM-UHFFFAOYSA-N
Synonym bromotriphenylmethane, triphenylmethyl bromide, trityl bromide, triphenylbromomethane, bromodiphenylmethyl benzene, methane, bromotriphenyl, benzene, 1,1',1-bromomethylidyne tris, triphenylmethylbromide, bromomethanetriyl tribenzene, 2-bromo-1,1,1-triphenylethane
PubChem CID 11692
IUPAC Name [bromo(diphenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br

Specifications

Melting Point 152°C to 156°C
Density 1.55
Boiling Point 230°C (15 mmHg)
Quantity 500 g
UN Number UN1759
Beilstein 1878494
Sensitivity Moisture sensitive
Formula Weight 323.24
Percent Purity 98%
Chemical Name or Material Bromotriphenylmethane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'