missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bronze Gauze, Alloy 220, 40 mesh woven from 0.25mm (0.01 in.) in dia. wire, Thermo Scientific™
Typical composition of alloy: Cu 89.0-91.0%, Fe 0.05% max, Pb 0.5% max Zn remainder
£34.20
Specifications
Chemical Name or Material | Bronze |
---|---|
Appearance | Bronze colored gauze |
MDL Number | MFCD00198187 |
Mesh Size | 220,40 Mesh |
Odor | Odorless |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15476795
|
Thermo Scientific Alfa Aesar
045166.HB |
300 x 300 mm | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Bronze | |
MFCD00198187 | |
Odorless |
Bronze colored gauze | |
220,40 Mesh | |
Gauze |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures