missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Class A Volumetric Flasks, Amber Glass
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks, Amber Glass are manufactured from durable DURAN™ glass tubing.
£67.80 - £125.00
Specifications
Certifications/Compliance | DIN EN ISO 1042 |
---|---|
Closure Color | Brown |
Color | Brown |
Class | A |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
11859050
|
Hirschmann™
2640280 |
100 mL | 12/21 NS Standard Ground |
£72.30
Pack of 2 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
||||
10116590
|
Hirschmann™
2640293 |
1000 mL | 24/29 NS Standard Ground |
£96.25
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
||||
11786722
|
Hirschmann™
2640281 |
100 mL | 14/23 NS Standard Ground |
£81.30
Pack of 2 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
||||
11878891
|
Hirschmann™
2640261 |
10 mL | 10/19 NS Standard Ground |
£107.00
Pack of 2 |
Estimated Shipment: 01-04-2025
Log in to see stock. |
||||
10761041
|
Hirschmann™
2640294 |
2000 mL | 29/32 NS Standard Ground |
£125.00
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
11756722
|
Hirschmann™
2640260 |
10 mL | 7/16 NS Standard Ground |
£70.90
Pack of 2 |
Estimated Shipment: 09-01-2025
Log in to see stock. |
||||
11766722
|
Hirschmann™
2640265 |
20 mL | 10/19 NS Standard Ground |
£69.90
Pack of 2 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
11776722
|
Hirschmann™
2640270 |
25 mL | 10/19 NS Standard Ground |
£108.00
Pack of 2 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
||||
11716742
|
Hirschmann™
2640285 |
200 mL | 14/23 NS Standard Ground |
£108.00
Pack of 2 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
||||
11682229
|
Hirschmann™
2640266 |
20 mL | 12/21 NS Standard Ground |
£67.80
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11736742
|
Hirschmann™
2640290 |
500 mL | 19/26 NS Standard Ground |
£123.00
Pack of 2 |
In Stock
Log in to see stock. |
||||
11706742
|
Hirschmann™
2640275 |
50 mL | 12/21 NS Standard Ground |
£72.90
Pack of 2 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||||
11839600
|
Hirschmann™
2640278 |
50 mL | 14/23 NS Standard Ground |
£72.30
Pack of 2 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
13187939
|
Hirschmann™
2640253 |
5 mL | 10/19 NS Standard Ground |
£69.90
Pack of 2 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||||
30090420
|
Hirschmann™
2640283 |
150 mL | 14/23 NS Standard Ground |
£73.45
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- With NS ground and hollow amber glass stopper
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
DIN EN ISO 1042 | |
Brown | |
Brown | |
A |