missing translation for 'onlineSavingsMsg'
Learn More

Steris™ Browne™ Control™ Autoclave Indicator Valves

Product Code. p-8000452
Click to view available options
Color:
Black Spot
Green Spot
White Spot
Yellow Spot
Unit Size:
Pack of 100

Product Code. 12715335

Brand: Steris™ 7302AB

Only null left
Add to basket
Edge
Add to basket

Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.

  • The use of TST Control™ emulating Indicators is recommended for monitoring porous load steam sterilization processes
  • Various colors
  • Glass tubes
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow Spot
For Use With (Application) For monitoring porous load steam sterilization processes
For Use With (Equipment) Autoclave
Product Type Browne Control Tube
Temperature (Metric) 134°C
Quantity 100 Pk.
Sterilization Indicator Yes
Sterilization Time 3 min.
Sterilization Method Steam, Dry Heat
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.