missing translation for 'onlineSavingsMsg'
Learn More

Brucine, 99%, anhydrous

Product Code. 10297303 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
50g
5g
This item is not returnable. View return policy

Product Code. 10297303

Brand: Thermo Scientific Acros 278725000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 357-57-3 | C23H26N2O4 | 394.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357-57-3
Molecular Formula C23H26N2O4
Molecular Weight (g/mol) 394.46
MDL Number MFCD00005942
InChI Key RRKTZKIUPZVBMF-VLTSECPOSA-N
Synonym brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy
PubChem CID 51413923
IUPAC Name (4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
SMILES COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC

Specifications

Melting Point 174.0°C to 179.0°C
Color White to Beige
Loss on Drying 1% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 500 g
Beilstein 27, IV, 7876
Merck Index 15, 1464
Specific Rotation Condition −123° (20°C c=2.5,CCl3)
Specific Rotation − 123.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, chloroform and benzene, slightly soluble in ether, glycerol and, ethyl acetate
Formula Weight 394.46
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Brucine, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado