missing translation for 'onlineSavingsMsg'
Learn More

Brucine dihydrate, 99%, Thermo Scientific Chemicals

Product Code. 10782391
100 g, Glass bottle
Click to view available options
Quantity:
50 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145428-94-0
Molecular Formula C23H27N2O4
Molecular Weight (g/mol) 395.48
MDL Number MFCD00005942
InChI Key RRKTZKIUPZVBMF-IBTVXLQLSA-O
Synonym brucine dihydrate, 10,11-dimethoxystrychnine, 10,11-dimethoxystrychnine dihydrate
PubChem CID 69897601
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;dihydrateShow MoreShow Less
SMILES COC1=CC2=C(C=C1OC)[C@@]13CC[NH+]4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34

Specifications

Melting Point 174°C to 179°C
Color White to Beige
Loss on Drying 6.0% to 9.5% (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Merck Index 14, 1455
Specific Rotation -123
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform and benzene,slightly soluble in ether,glycerol and,ethyl acetate
Formula Weight 430.5
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material Brucine dihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Brucine dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brucine dihydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.