missing translation for 'onlineSavingsMsg'
Learn More
Brucine sulfate hydrate, 98% (dry wt.), water <13%, Thermo Scientific Chemicals Product Code.: 11436390

Brucine sulfate hydrate, 98% (dry wt.), water <13%, Thermo Scientific Chemicals

Product Code. 11436390
100 g
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11436390

Brand: Thermo Scientific Alfa Aesar A19943.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652154-10-4
Molecular Formula C46H54N4O12S
Molecular Weight (g/mol) 887.01
MDL Number MFCD00013472
InChI Key HCMSIGALSOEZRW-MDQLRNFDSA-N
Synonym brucine sulfate salt
PubChem CID 118797651
IUPAC Name 10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;hydrateShow MoreShow Less
SMILES OS(O)(=O)=O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1Show MoreShow Less

Specifications

Melting Point 191°C to 196°C (decomposition)
Quantity 100 g
UN Number UN2811
Sensitivity Hygroscopic
Merck Index 14,1455
Solubility Information Soluble in water.
Formula Weight 443.52 (Anhydrous)
Percent Purity 98%
Assay (dry wt.), water <13%
Chemical Name or Material Brucine sulfate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252400 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog NumberA19943.14,A19943.22
Lot Number10213089 Certificate TypeCertificate of Analysis Date06/09/2022 Catalog NumberA19943.14,A19943.22
Lot NumberH3853B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19943.14,A19943.22
Lot NumberH1463A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19943.14,A19943.22
Lot Number10163284 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19943.14,A19943.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brucine sulfate hydrate, 98% (dry wt.), water <13%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.