missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BS3 (bis(sulfosuccinimidyl)suberate)
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
8 x 2 mg
10 x 2 mg
1 g
Unit Size:
1g
50mg
Pack of 10
Pack of 8
Description
Thermo Scientific™ Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.
Highlights:
- Crosslink cell-surface proteins before cell lysis and immunoprecipitation
- Identify receptor-ligand interactions by chemical crosslinking
- “Fix” protein interactions to allow identification of weak or transient protein interactions
- Identify near-neighbor protein interactions
- Protein crosslinking to create bioconjugates via single-step reactions
- Immobilize proteins onto amine-coated surfaces
Specifications
Specifications
| Chemical Reactivity | Amine-Amine |
| Cleavable | No |
| Molecular Weight (g/mol) | 572.43 |
| PEGylated | No |
| Spacer Arm Length | 11.4 Å |
| Content And Storage | Upon receipt store desiccated at 4°C. |
| Cell Permeability | No |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Labeling Method | Chemical Labeling |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction