missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BS3 (bis(sulfosuccinimidyl)suberate)

Product Code. p-4530976 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
8 x 2 mg
10 x 2 mg
1 g
Unit Size:
1g
50mg
Pack of 10
Pack of 8
This item is not returnable. View return policy

Product Code. 11841245

Brand: Thermo Scientific™ 21585

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Crosslink amines to amines with BS3; composed of sulfo-NHS-ester groups separated by an 11.4A, water-soluble spacer arm that is useful for cell-surface protein crosslinking.

Thermo Scientific™ Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.

Highlights:

  • Crosslink cell-surface proteins before cell lysis and immunoprecipitation
  • Identify receptor-ligand interactions by chemical crosslinking
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Identify near-neighbor protein interactions
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-to-Amine
Cleavable No
Formulation Bis(sulfosuccinimidyl) suberate; solid
Membrane Permeable No
Molecular Weight (g/mol) 572.43
PEGylated No
Sufficient For 28mL of typical 1X (1mM) reaction solution (total), or 8 separate uses
Spacer Arm Length 11.4Å
Quantity 8 x 2 mg
Solubility Water
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.