missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent

Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.

£191.00 - £388.00

Specifications

Storage 2°C to 8°C
View More Specs

Products 5
Product Code Brand Standard Components Quantity Price Quantity & Availability  
Product Code Brand Standard Components Quantity Price Quantity & Availability  
11557851
View Documents Promotion Details
Thermo Scientific™
TS-38831
BSTFA +1% TMCS 10 x 1 mL ampules
£277.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11597570
View Documents Promotion Details
Thermo Scientific™
TS-38832
BSTFA +1% TMCS 10 g
£191.00
Each
Please to purchase this item. Need a web account? Register with us today!
11527851
View Documents Promotion Details
Thermo Scientific™
TS-38833
BSTFA +1% TMCS 25 g
£260.00
Each
Please to purchase this item. Need a web account? Register with us today!
11517871
View Documents Promotion Details
Thermo Scientific™
TS-38834
BSTFA +1% TMCS 100 g
£388.00
Each
Please to purchase this item. Need a web account? Register with us today!
11577841
View Documents
Thermo Scientific™
TS-38840
BSTFA +10% TMCS 10 x 1 mL Ampules
£305.91
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.

  • Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
  • Catalyzed formulation is stronger than BSTFA alone

Specifications

Specifications

2°C to 8°C
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.