Learn More
Thermo Scientific™ BSTFA Silylation Reagent
Shop All Thermo Scientific ProductsDescription
Obtain excellent GC separations with Thermo Scientific™ BSTFA Silylation Reagent.
Improve GC analysis with Thermo Scientific™ BSTFA Silylation Reagent. The highly volatile byproducts mono(trimethylsilyl)trifluoroacetamide (BSTFA) and trifluoroacetamide elute with the solvent front to ensure excellent chromatographic separations. This silylation reagent reacts with a wide range of polar compounds and is particularly well suited for the GC analyses of some lower boiling TMS-amino acids and TMS Krebs cycle acids.
- Increased volatility of reaction byproducts mono(trimethylsilyl)trifluoroacetamide and trifluoroacetamide over corresponding nonfluorinated compounds from BSA
- Increased volatility makes it possible to derivatize smaller molecules with which the TMS derivatives elute with the byproducts from BSA
Specifications
Specifications
| For Use With | GC |
| Standard Concentration | 98 to 100% |
| Item Description | 100 g |
| Storage | 2°C to 8°C |
| Quantity | 100 g |
| Standard Components | Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate |
Safety and Handling
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl) acetimidate
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.