missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BSTFA Silylation Reagent

Obtain excellent GC separations with Thermo Scientific™ BSTFA Silylation Reagent.

Brand:  Thermo Scientific™ TS-38830

141.86 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



 View more versions of this product

Product Code. 11776957

  • £206.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Improve GC analysis with Thermo Scientific™ BSTFA Silylation Reagent. The highly volatile byproducts mono(trimethylsilyl)trifluoroacetamide (BSTFA) and trifluoroacetamide elute with the solvent front to ensure excellent chromatographic separations. This silylation reagent reacts with a wide range of polar compounds and is particularly well suited for the GC analyses of some lower boiling TMS-amino acids and TMS Krebs cycle acids.

BSTFA is a powerful thrimethylsilyl donor, with donor strength that is comparable to its unfluorinated analog BSA [N,O-Bis(trimethylsilyl)acetamide]. BSTFA reacts to replace labile hydrogens on a wide range of polar compounds with a -Si(CH3)3 group. This physical characteristic is particularly useful in the gas chromatography of some lower boiling TMS-amino acids and TMS Krebs cycle acids.
  • Increased volatility of reaction byproducts mono(trimethylsilyl)trifluoroacetamide and trifluoroacetamide over corresponding nonfluorinated compounds from BSA
  • Increased volatility makes it possible to derivatize smaller molecules with which the TMS derivatives elute with the byproducts from BSA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC
10 x 1 mL ampules
10 x 1 mL ampules
98 to 100%
2°C to 8°C
Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    27

    Hours

    1

    Minutes

    37

    Seconds

    18
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

    Days

    58

    Hours

    1

    Minutes

    37

    Seconds

    18
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ BSTFA Silylation Reagent > 10 x 1mL ampules

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.