Learn More
Corning™ BTD Valve Head Complete
Corning™ valve head complete
Brand:  Corning™ 6851
 View more versions of this product
Product Code. 16802100
  • £94.96 / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
BTD valve head complete
2.5 to 10 mL
Dispensing Organic or Inorganic Liquids