missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA, Rat anti-Human, Clone: 6F4, Abnova™
Rat Monoclonal Antibody
Brand: Abnova MAB16112.100ug
This item is not returnable.
View return policy
Specifications
| BTLA | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10% glycerol). | |
| BTLA1/CD272/FLJ16065/MGC129743 | |
| Rat | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen) | |
| 6F4 | |
| Immunohistochemistry (Frozen sections) The optimal working dilution should be determined by the end user. | |
| B and T lymphocyte associated | |
| BTLA | |
| Ectodomain of human BTLA fused to human IgG Fc. | |
| RUO | |
| 151888 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction