missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN2A2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547670
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant mouse (rm) BTN and rmBTNL2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
BTN2A2 Polyclonal antibody specifically detects BTN2A2 in Mouse samples. It is validated for Western Blot.Specifications
BTN2A2 | |
Polyclonal | |
Unconjugated | |
BTN2A2 | |
BT2.2; BTF2; Btn2; BTN2.2; BTN2A2; butyrophilin 2; butyrophilin subfamily 2 member A2; butyrophilin, subfamily 2, member A2; D030063K05 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
238555 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
A4QPC6 | |
BTN2A2 | |
Mouse myeloma cell line NS0-derived recombinant mouse BTN2A2/Butyrophilin 2 Gln29-Ser219 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |