missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BTT 3033
Selective inhibitor of integrin α2β1
£248.00 - £965.00
Specifications
Purity | >98% |
---|---|
Product Type | BTT 3033 |
Molecular Weight (g/mol) | 465.5 |
Inhibitors | BTT 3033 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18738599
|
Tocris Bioscience™
4724/10 |
10 mg |
£248.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18733158
|
Tocris Bioscience™
4724/50 |
50 mg |
£965.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective inhibitor of integrin α2β1 (EC50 = 130 nM for α2β1 binding to collagen I). Binds to the α2I domain. Exhibits selectivity for α2β1 over integrins α3β1, α4β1, α5β1 and αv. Inhibits platelet aggregation to collagen I coated capillaries under flow. Also inhibits binding of α2-expressing CHO cells to collagen I under shear stress conditions.Specifications
>98% | |
BTT 3033 | |
465.5 | |
BTT 3033 |
For Research Use Only